Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBP7
Entrez Gene ID
838780
Gene Name
F-box protein 7
Gene Aliases
AT1G21760, ATFBP7, F-BOX PROTEIN 7, F-box protein 7, F8K7.20, F8K7_20
Chromosome Location
Chr.1: 7649194 - 7651816 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332527.1 NP_001320240.1 8 - 9 1028 103
NM_102025.3 NP_564150.1 8 - 9 840 103
GenBank mRNA AK228033.1 - 8 - 9 760 103
BT006427.1 - 7 - 8 710 103

View Full Product Details