Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GC5
Entrez Gene ID
844322
Gene Name
golgin Putative 5
Gene Aliases
AT1G79830, F19K16.21, F19K16_21, golgin candidate 5
Chromosome Location
Chr.1: 30027569 - 30033907 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001124150.1 NP_001117622.1 14 - 15 2435 66
NM_001198512.1 NP_001185441.1 14 - 15 2608 66
NM_001198513.2 NP_001185442.1 14 - 15 2548 66
NM_001334925.1 NP_001321438.1 14 - 15 2554 66
NM_106632.4 NP_178101.3 14 - 15 2535 66
GenBank mRNA AK226727.1 - 14 - 15 2437 66
EU249329.1 - 13 - 14 2278 66

View Full Product Details