Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCA2;4
Entrez Gene ID
844377
Gene Name
Cyclin A2;4
Gene Aliases
AT1G80370, Cyclin A2;4, F5I6.12, F5I6_12
Chromosome Location
Chr.1: 30213705 - 30217102 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106686.3 NP_178153.1 4 - 5 830 101

View Full Product Details