Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CARA
Entrez Gene ID
822396
Gene Name
carbamoyl phosphate synthetase A
Gene Aliases
AT3G27740, CARBAMOYL PHOSPHATE SYNTHETASE SMALL SUBUNIT, VEN6, VENOSA 6, carbamoyl phosphate synthetase A
Chromosome Location
Chr.3: 10281230 - 10283935 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084749.1 NP_001078218.1 2 - 3 352 79
NM_113690.3 NP_566824.1 2 - 3 421 79
GenBank mRNA AY046004.1 - 2 - 3 352 79
AY079315.1 - 2 - 3 278 79

View Full Product Details