Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCA2
Entrez Gene ID
823927
Gene Name
ATP-binding cassette A2
Gene Aliases
A. THALIANA ABC2 HOMOLOG 1, ABC2 homolog 1, AT3G47730, ATATH1, ATH1, ATP-binding cassette A2
Chromosome Location
Chr.3: 17594060 - 17599083 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114641.4 NP_190357.2 7 - 8 1675 75
GenBank mRNA BT004197.1 - 7 - 8 1488 75
BT005456.1 - 7 - 8 1420 75

View Full Product Details