Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOPP7
Entrez Gene ID
826726
Gene Name
Calcineurin-like metallo-phosphoesterase superfamily protein
Gene Aliases
AT4G11240, F8L21.30, F8L21_30
Chromosome Location
Chr.4: 6847112 - 6849317 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117195.5 NP_567375.1 3 - 4 1176 116
GenBank mRNA AY086060.1 - 3 - 4 1079 116
AY090365.1 - 3 - 4 1123 116

View Full Product Details