Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD51
Entrez Gene ID
832208
Gene Name
RAS associated with diabetes protein 51
Gene Aliases
AT5G20850, ATRAD51, F22D1.20, F22D1_20, RAS associated with diabetes protein 51
Chromosome Location
Chr.5: 7070491 - 7072968 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343688.1 NP_001332342.1 2 - 3 349 73
NM_122092.3 NP_568402.1 2 - 3 347 73
GenBank mRNA U43528.1 - 2 - 3 291 73

View Full Product Details