Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSLA02
Entrez Gene ID
832337
Gene Name
cellulose synthase-like A02
Gene Aliases
ARABIDOPSIS THALIANA CELLULOSE SYNTHASE-LIKE A02, ARABIDOPSIS THALIANA CELLULOSE SYNTHASE-LIKE A2, ARABIDOPSIS THALIANA CELLULOSE SYNTHASE-LIKE A02, ARABIDOPSIS THALIANA CELLULOSE SYNTHASE-LIKE A2, AT5G22740, ATCSLA02, ATCSLA2, CELLULOSE SYNTHASE-LIKE A 2, CSLA2, MDJ22.16, MDJ22_16, cellulose synthase-like A02
Chromosome Location
Chr.5: 7554689 - 7560190 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122180.3 NP_197666.1 8 - 9 1765 93
GenBank mRNA AY058158.1 - 8 - 9 1648 93
AY059852.1 - 8 - 9 1641 93
AY093293.1 - 8 - 9 1441 93
KJ138672.1 - 8 - 9 1441 93

View Full Product Details