Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CHX3
- Entrez Gene ID
-
832354
- Gene Name
- cation/H+ exchanger 3
- Gene Aliases
-
ARABIDOPSIS THALIANA CATION/H+ EXCHANGER 3, AT5G22900, ATCHX3, MRN17.13, MRN17_13, cation/H+ exchanger 3
- Chromosome Location
-
Chr.5: 7657224 - 7659868 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122196.1 |
NP_197681.1 |
1 - 2 |
269 |
83 |
View Full Product Details