Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDPK9
Entrez Gene ID
832424
Gene Name
calmodulin-like domain protein kinase 9
Gene Aliases
ARABIDOPSIS THALIANA CALMODULIN-LIKE DOMAIN PROTEIN KINASE 9, AT5G23580, ATCDPK9, ATCPK12, CALCIUM-DEPENDENT PROTEIN KINASE, CALCIUM-DEPENDENT PROTEIN KINASE 12, CPK12, calmodulin-like domain protein kinase 9
Chromosome Location
Chr.5: 7950202 - 7952619 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122264.5 NP_197748.1 2 - 3 858 123
GenBank mRNA AK229078.1 - 2 - 3 731 123
BT026382.1 - 2 - 3 672 123
U20388.1 - 2 - 3 709 123

View Full Product Details