Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATSAC1
Entrez Gene ID
838868
Gene Name
Phosphoinositide phosphatase family protein
Gene Aliases
AT1G22620, F12K8.3, SAC1, suppressor of actin 1
Chromosome Location
Chr.1: 7994166 - 8002989 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102109.4 NP_173676.2 8 - 9 1428 123
GenBank mRNA AF266460.1 - 8 - 9 1226 123
AY227244.1 - 8 - 9 1226 123

View Full Product Details