Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGT74B1
Entrez Gene ID
839022
Gene Name
UDP-glucosyl transferase 74B1
Gene Aliases
AT1G24100, F3I6.2, F3I6_2, UDP-glucosyl transferase 74B1
Chromosome Location
Chr.1: 8525334 - 8527087 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102256.3 NP_173820.1 1 - 2 738 131
GenBank mRNA AF387008.1 - 1 - 2 739 131
BT001160.1 - 1 - 2 661 131
KJ138987.1 - 1 - 2 661 131

View Full Product Details