Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CAL
- Entrez Gene ID
-
839172
- Gene Name
- K-box region and MADS-box transcription factor family protein
- Gene Aliases
-
AGAMOUS-like 10, AGL10, AT1G26310, CAL1, CAULIFLOWER, F28B23.25, F28B23_25, MADS3
- Chromosome Location
-
Chr.1: 9099994 - 9103644 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102395.3 |
NP_564243.1 |
2 - 3 |
402 |
68 |
GenBank mRNA |
AB493477.1 |
- |
2 - 3 |
268 |
68 |
View Full Product Details