Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESP1
Entrez Gene ID
843720
Gene Name
hydroxyproline-rich glycoprotein family protein
Gene Aliases
AT1G73840, ENHANCED SILENCING PHENOTYPE 1, F25P22.26, F25P22_26
Chromosome Location
Chr.1: 27763617 - 27766372 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106044.6 NP_177525.2 2 - 3 222 57
GenBank mRNA AY070424.1 - 2 - 3 214 57
AY096602.1 - 2 - 3 178 57

View Full Product Details