Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YSL4
Entrez Gene ID
834102
Gene Name
YELLOW STRIPE like 4
Gene Aliases
AT5G41000, AtYSL4, MEE6.7, MEE6_7, YELLOW STRIPE like 4
Chromosome Location
Chr.5: 16420755 - 16423830 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344377.1 NP_001331152.1 2 - 3 679 77
NM_123465.4 NP_198916.2 2 - 3 679 77
GenBank mRNA AY515562.1 - 2 - 3 524 77

View Full Product Details