Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DUR3
- Entrez Gene ID
-
834574
- Gene Name
- urea-proton symporter DEGRADATION OF UREA 3 (DUR3)
- Gene Aliases
-
AT5G45380, ATDUR3, DEGRADATION OF UREA 3, MFC19.5, MFC19_5
- Chromosome Location
-
Chr.5: 18390942 - 18395928 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123906.5 |
NP_199351.2 |
5 - 6 |
1077 |
87 |
View Full Product Details