Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LTI65
Entrez Gene ID
835306
Gene Name
CAP160 protein
Gene Aliases
AT5G52300, K24M7.3, K24M7_3, LOW-TEMPERATURE-INDUCED 65, RD29B, RESPONSIVE TO DESSICATION 29B
Chromosome Location
Chr.5: 21237021 - 21239531 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036984.1 NP_001032061.1 4 - 5 1923 66
NM_124609.4 NP_200043.2 4 - 5 1961 66
GenBank mRNA AY081282.1 - 4 - 5 1925 66
AY128731.1 - 4 - 5 1777 66

View Full Product Details