Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VLN2
Entrez Gene ID
818773
Gene Name
villin 2
Gene Aliases
AT2G41740, ATVLN2, T11A7.16, T11A7_16, villin 2
Chromosome Location
Chr.2: 17410672 - 17417820 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336936.1 NP_001323674.1 22 - 23 3271 78
NM_129738.5 NP_565958.1 22 - 23 3041 78
GenBank mRNA AK226882.1 - 22 - 23 2736 78
AY080601.1 - 22 - 23 2801 78
AY133782.1 - 21 - 22 2598 78

View Full Product Details