Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCC15
- Entrez Gene ID
-
825269
- Gene Name
- multidrug resistance-associated protein 15
- Gene Aliases
-
AT3G60970, ATMRP15, ATP-binding cassette C15, MRP15, multidrug resistance-associated protein 15
- Chromosome Location
-
Chr.3: 22557282 - 22561716 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115961.3 |
NP_191656.2 |
8 - 9 |
2981 |
139 |
View Full Product Details