Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP94D2
- Entrez Gene ID
-
824830
- Gene Name
- cytochrome P450, family 94, subfamily D, polypeptide 2
- Gene Aliases
-
''cytochrome P450, AT3G56630, cytochrome P450, family 94, polypeptide 2, polypeptide 2'', subfamily D
- Chromosome Location
-
Chr.3: 20978901 - 20980772 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115522.4 |
NP_191222.1 |
2 - 2 |
1598 |
138 |
View Full Product Details