Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PAI2
- Entrez Gene ID
-
830442
- Gene Name
- phosphoribosylanthranilate isomerase 2
- Gene Aliases
-
AT5G05590, MOP10.13, MOP10_13, PHOSPHORIBOSYLANTHRANILATE ISOMERASE, phosphoribosylanthranilate isomerase 2
- Chromosome Location
-
Chr.5: 1666471 - 1668569 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203005.2 |
NP_974734.1 |
3 - 3 |
654 |
143 |
View Full Product Details