Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP1R1B
Entrez Gene ID
282459
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 1B
Gene Aliases
-
Chromosome Location
Chr.19: 40006493 - 40015037 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174647.2 NP_777072.1 6 - 7 897 96
GenBank mRNA BC118264.1 - 6 - 7 632 96
M27444.1 - 6 - 7 897 96

View Full Product Details