Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CATHL1
- Entrez Gene ID
-
282164
- Gene Name
- cathelicidin 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 51579582 - 51581008 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174825.1 |
NP_777250.1 |
1 - 2 |
210 |
113 |
GenBank mRNA |
BC114189.1 |
- |
1 - 2 |
212 |
113 |
- Gene Symbol
- LOC112441458
- Entrez Gene ID
-
112441458
- Gene Name
- cathelicidin-1-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 51599665 - 51601123 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC142024.1 |
- |
1 - 2 |
215 |
113 |
L08834.1 |
- |
1 - 2 |
210 |
113 |
View Full Product Details