Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GKN1
Entrez Gene ID
407211
Gene Name
gastrokine 1
Gene Aliases
FOV
Chromosome Location
Chr.11: 67308836 - 67314682 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001148.1 NP_001001148.1 5 - 6 504 73
GenBank mRNA AY456959.1 - 5 - 6 504 73

View Full Product Details