Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPLP1
Entrez Gene ID
533285
Gene Name
ribosomal protein lateral stalk subunit P1
Gene Aliases
-
Chromosome Location
Chr.10: 16343828 - 16345976 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025340.2 NP_001020511.1 3 - 4 396 93
GenBank mRNA AY911313.1 - 3 - 4 397 93
BC102695.1 - 1 - 1 398 93

View Full Product Details