Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YIPF5
Entrez Gene ID
521829
Gene Name
Yip1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.7: 55257073 - 55275266 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034543.1 NP_001029715.1 4 - 5 556 110
GenBank mRNA BC102241.1 - 4 - 5 556 110
BT020972.1 - 4 - 5 517 110
BT029906.1 - 4 - 5 494 110

View Full Product Details