Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX39A
Entrez Gene ID
614215
Gene Name
DExD-box helicase 39A
Gene Aliases
DDX39
Chromosome Location
Chr.7: 11344663 - 11353461 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034752.1 NP_001029924.1 3 - 4 439 96
GenBank mRNA BC104616.1 - 3 - 4 392 96
BT021050.1 - 3 - 4 439 96

View Full Product Details