Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDX39A
Entrez Gene ID
614215
Gene Name
DExD-box helicase 39A
Gene Aliases
DDX39
Chromosome Location
Chr.7: 11344663 - 11353461 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034752.1 NP_001029924.1 9 - 10 1222 70
GenBank mRNA BC104616.1 - 10 - 11 1236 70
BT021050.1 - 9 - 10 1222 70

View Full Product Details