Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHDC8B
Entrez Gene ID
527321
Gene Name
kelch domain containing 8B
Gene Aliases
-
Chromosome Location
Chr.22: 50843123 - 50863829 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035370.2 NP_001030447.2 4 - 5 960 92
GenBank mRNA BT020815.1 - 4 - 5 960 92
BT026296.1 - 4 - 5 950 92

View Full Product Details