Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRPG
Entrez Gene ID
522621
Gene Name
small nuclear ribonucleoprotein polypeptide G
Gene Aliases
-
Chromosome Location
Chr.11: 68602486 - 68611488 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040543.3 NP_001035633.1 1 - 2 119 92
GenBank mRNA BC103236.1 - 1 - 1 106 92
CB426020.1 - 1 - 2 87 92
EV659942.1 - 1 - 2 123 92

View Full Product Details