Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CABP4
Entrez Gene ID
281656
Gene Name
calcium binding protein 4
Gene Aliases
-
Chromosome Location
Chr.29: 45343481 - 45346978 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174256.3 NP_776681.1 2 - 3 458 75
GenBank mRNA AY048883.1 - 2 - 3 459 75

View Full Product Details