Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRA1
Entrez Gene ID
281886
Gene Name
killer cell lectin-like receptor subfamily A, member 1
Gene Aliases
Ly49
Chromosome Location
Chr.5: 98969285 - 98984168 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174376.2 NP_776801.1 3 - 4 343 106
GenBank mRNA AY075101.1 - 3 - 4 343 106

View Full Product Details