Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTSF1
Entrez Gene ID
506864
Gene Name
gametocyte specific factor 1
Gene Aliases
FAM112B
Chromosome Location
Chr.5: 25613240 - 25632602 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034273.2 NP_001029445.1 4 - 5 358 100
GenBank mRNA BC102713.1 - 4 - 5 359 100

View Full Product Details