Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN11
Entrez Gene ID
508268
Gene Name
claudin 11
Gene Aliases
-
Chromosome Location
Chr.1: 96999959 - 97014887 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035055.2 NP_001030227.1 1 - 2 430 73
GenBank mRNA BC105204.1 - 1 - 2 431 73

View Full Product Details