Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDF1
Entrez Gene ID
515380
Gene Name
endothelial differentiation related factor 1
Gene Aliases
-
Chromosome Location
Chr.11: 105948291 - 105952197 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035307.2 NP_001030384.1 3 - 4 384 68
GenBank mRNA BC102246.1 - 3 - 4 386 68

View Full Product Details