Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSPAN3
Entrez Gene ID
616881
Gene Name
tetraspanin 3
Gene Aliases
-
Chromosome Location
Chr.21: 32177529 - 32204399 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035487.1 NP_001030564.1 4 - 5 551 133
GenBank mRNA BC102734.1 - 4 - 5 551 133

View Full Product Details