Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFDN4
Entrez Gene ID
514621
Gene Name
prefoldin subunit 4
Gene Aliases
-
Chromosome Location
Chr.13: 81632576 - 81644895 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038540.1 NP_001033629.1 2 - 3 183 92
GenBank mRNA BC109756.1 - 2 - 3 183 92

View Full Product Details