Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLIPR1
Entrez Gene ID
767905
Gene Name
GLI pathogenesis related 1
Gene Aliases
-
Chromosome Location
Chr.5: 4964662 - 5034037 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076984.2 NP_001070452.1 4 - 5 718 91
GenBank mRNA BC102295.1 - 4 - 5 720 91

View Full Product Details