Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFB2M
Entrez Gene ID
516874
Gene Name
transcription factor B2, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.16: 30802365 - 30820745 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038127.2 NP_001033216.1 7 - 8 1162 130
GenBank mRNA BC109634.1 - 7 - 8 1163 130

View Full Product Details