Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUPT20H
Entrez Gene ID
534126
Gene Name
SPT20 homolog, SAGA complex component
Gene Aliases
FAM48A
Chromosome Location
Chr.12: 24646264 - 24685744 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046370.1 NP_001039835.1 17 - 18 1469 117
GenBank mRNA BC105571.1 - 17 - 18 1469 117

View Full Product Details