Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD83
Entrez Gene ID
617034
Gene Name
CD83 molecule
Gene Aliases
-
Chromosome Location
Chr.23: 42655919 - 42676052 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046590.1 NP_001040055.1 1 - 2 149 98
GenBank mRNA BC112861.1 - 1 - 2 149 98

View Full Product Details