Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFPL1
Entrez Gene ID
613442
Gene Name
zinc finger protein like 1
Gene Aliases
-
Chromosome Location
Chr.29: 43297356 - 43301446 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076272.1 NP_001069740.1 2 - 3 266 62
GenBank mRNA BC110275.1 - 2 - 3 266 62
BT025470.1 - 2 - 3 296 62

View Full Product Details