Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIG1
Entrez Gene ID
614409
Gene Name
androgen induced 1
Gene Aliases
-
Chromosome Location
Chr.9: 80508387 - 80778252 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076326.1 NP_001069794.1 2 - 3 327 121
GenBank mRNA BC112809.1 - 2 - 3 327 121

View Full Product Details