Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESAM
Entrez Gene ID
616926
Gene Name
endothelial cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.29: 28232745 - 28241470 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078066.2 NP_001071534.1 6 - 7 1029 66
GenBank mRNA BC105434.1 - 6 - 7 1030 66

View Full Product Details