Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK7
Entrez Gene ID
506380
Gene Name
kallikrein related peptidase 7
Gene Aliases
KLND
Chromosome Location
Chr.18: 56960737 - 56965958 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075256.1 NP_001068724.1 4 - 5 584 94
GenBank mRNA BC123882.1 - 4 - 5 584 94

View Full Product Details