Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFA2C
Entrez Gene ID
618389
Gene Name
BPI fold containing family A, member 2C
Gene Aliases
BSP30C
Chromosome Location
Chr.13: 62565063 - 62575212 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076503.1 NP_001069971.1 4 - 5 673 97
GenBank mRNA DQ835286.1 - 4 - 5 673 97

View Full Product Details