Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTPS1
Entrez Gene ID
506131
Gene Name
CTP synthase 1
Gene Aliases
CTPS
Chromosome Location
Chr.3: 105201090 - 105232559 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077858.2 NP_001071326.1 15 - 16 1561 96
GenBank mRNA BC126649.1 - 15 - 16 1563 96

View Full Product Details