Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRMS1L
Entrez Gene ID
514432
Gene Name
breast cancer metastasis-suppressor 1 like
Gene Aliases
-
Chromosome Location
Chr.21: 46014187 - 46058396 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083428.1 NP_001076897.1 8 - 9 748 83
GenBank mRNA BC123661.1 - 8 - 9 748 83

View Full Product Details