Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TOR2A
Entrez Gene ID
534311
Gene Name
torsin family 2 member A
Gene Aliases
Prosalusin, Torsin-2A
Chromosome Location
Chr.11: 98411720 - 98416236 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083476.2 NP_001076945.1 4 - 5 747 68
GenBank mRNA BC114885.1 - 4 - 5 748 68

View Full Product Details