Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JAM2
Entrez Gene ID
538846
Gene Name
junctional adhesion molecule 2
Gene Aliases
-
Chromosome Location
Chr.1: 10704997 - 10787558 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083736.1 NP_001077205.1 5 - 6 852 93
GenBank mRNA BC134600.1 - 5 - 6 852 93

View Full Product Details